Learn More
Cayman Chemical 24-dehydro Cholesterol, Purity: ≥98%, CAS: 313-04-2, 10 mg
SDP
Catalog No. 502970105
Encompass_Preferred
An immediate precursor to cholesterol in the Bloch pathway of cholesterol biosynthesis; reported to activate liver X receptor-target genes in both the liver of cholesterol-free mice and in cholesterol-starved macrophage foam cells in atherosclerotic lesions
Catalog No. 50-297-0105 Supplier Cayman Chemical Supplier No. 1494310 MG
An immediate precursor to cholesterol in the Bloch pathway of cholesterol biosynthesis; reported to activate liver X receptor-target genes in both the liver of cholesterol-free mice and in cholesterol-starved macrophage foam cells in atherosclerotic lesions