Learn More
eMolecules​ 307496-26-0 | 2,4-Difluorobenzylzinc bromide, 0.50 M in THF | Synthonix | MFCD01311423 | 272.400 | C7H5BrF2Zn | 0.000 | [Br-].Fc1ccc(C[Zn+])c(F)c1 | 25ml | 627353597
Catalog No. 502695369 Shop All eMolecules Inc. Products
Encompass_Preferred
2,4-Difluorobenzylzinc bromide, 0.50 M in THF | Synthonix | 307496-26-0 | MFCD01311423 | 272.400 | C7H5BrF2Zn | 0.000 | [Br-].Fc1ccc(C[Zn+])c(F)c1 | 25ml | 627353597
Catalog No. 50-269-5369 Supplier eMolecules​ Supplier No. 627353597
2,4-Difluorobenzylzinc bromide, 0.50 M in THF | Synthonix | 307496-26-0 | MFCD01311423 | 272.400 | C7H5BrF2Zn | 0.000 | [Br-].Fc1ccc(C[Zn+])c(F)c1 | 25ml | 627353597