Learn More
Bioworld 24-Epibrassinolide, 100 mg
Catalog No. 50255033
Encompass_Preferred
24-EpibrassinolideSynonym: (22R, 23R, 24R)-2α, 3α, 22, 23-Tetrahydroxy-24-methyl-β-homo-7-oxa-5-cholestan-6-one Inhibits aging, break dormancy, stimulates germination, promotes growth of foliage plant, and induces hermaphrodite flower and female flowerAppearance: White crystals Melting Point: 254-256°C Soluble in DMSO
Catalog No. 50-255-033 Supplier Bioworld Supplier No. 306310031
24-EpibrassinolideSynonym: (22R, 23R, 24R)-2α, 3α, 22, 23-Tetrahydroxy-24-methyl-β-homo-7-oxa-5-cholestan-6-one Inhibits aging, break dormancy, stimulates germination, promotes growth of foliage plant, and induces hermaphrodite flower and female flowerAppearance: White crystals Melting Point: 254-256°C Soluble in DMSO