Learn More
Hach Company 24 Glass Bottles 350mL SD900
Supplier:  Hach Company RF240350
Encompass
Kit-24 Glass Bottles-350mL-for SD900 Refrigerated SamplerRefrigerated bottle kit with (24) 350 mL glass bottles with caps retainer and full bottle shut-off
Catalog No. 50-002-07256