Learn More
Alkali Scientific ASI™ 24-Hour Urine 3.5L Containers, Leak-Resistant | Pathology, Microbiology, Histology,40/Cs, Vertical Space Saver, With EZ-UP Pour-Spout
SDP
Catalog No. 503026603
Encompass_Preferred
Alkali Scientific™ 24hr Urine Collection and Transport Containers are feature packed, user friendly with unparallel value and space saver styles for all storage formats. Ergonomic Design Balanced CG Capacity: 3.5L EZ-UP Spout Cap (optional) Manufactured from pure virgin, UV filtering, medical grade HDPE Choice of integrated 30mL 6N HCl or none (Value style)
Catalog No. 50-302-6603 Supplier Alkali Scientific Supplier No. 24UC013
Alkali Scientific™ 24hr Urine Collection and Transport Containers are feature packed, user friendly with unparallel value and space saver styles for all storage formats. Ergonomic Design Balanced CG Capacity: 3.5L EZ-UP Spout Cap (optional) Manufactured from pure virgin, UV filtering, medical grade HDPE Choice of integrated 30mL 6N HCl or none (Value style)