Learn More
Sentry Air Systems 24 in Ducted Fume Hood
SDP
Catalog No. NC2885521
Encompass
The 24 in. wide Ducted Fume Hood provides an external ducting solution for chemical laboratories and applications that use large amounts of volatile chemicals. This exhaust fume hood uses a powerful fan (not included) to draw fumes through the ductwork (not included) to release outside of the building. This system can be used for applications such as evaporating large amounts of volatile organics, working with chemicals that are hard to filter or have a low odor threshold, and heavy powder filling. This hood is constructed of all chemical resistant materials and includes a 4 exhaust collar. This compact and versatile exhaust hood offers an economical solution for applications that require external ducting. Please Note: The end user is responsible for conforming to local, state, and federal guidelines for external ducting. This exhaust fume hood does not come with a fan, external ductwork, or a filter.
Catalog No. NC2885521 Supplier Sentry Air Systems Supplier No. SS324E
The 24 in. wide Ducted Fume Hood provides an external ducting solution for chemical laboratories and applications that use large amounts of volatile chemicals. This exhaust fume hood uses a powerful fan (not included) to draw fumes through the ductwork (not included) to release outside of the building. This system can be used for applications such as evaporating large amounts of volatile organics, working with chemicals that are hard to filter or have a low odor threshold, and heavy powder filling. This hood is constructed of all chemical resistant materials and includes a 4 exhaust collar. This compact and versatile exhaust hood offers an economical solution for applications that require external ducting. Please Note: The end user is responsible for conforming to local, state, and federal guidelines for external ducting. This exhaust fume hood does not come with a fan, external ductwork, or a filter.