Learn More
Medchemexpress LLC HY-101737 5mg Medchemexpress, 24-Norursodeoxycholic acid CAS:99697-24-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1017375MG
Encompass_Preferred
Medchemexpress, HY-101737 5mg 24-Norursodeoxycholic acid CAS:99697-24-2 24-norursodeoxycholic acid (norUDCA) is a side chain-shortened C23 homologue of UDCA and has shown potent anti-cholestatic, anti-inflammatory and anti-fibrotic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7866