Learn More
USA Scientific Inc 24-box (3 x 4) upright freezer rack for 50-place boxes
Catalog No. 507541195
Encompass
24-box (3 long x 4 high) upright freezer rack for 50-place boxes, 19 in L x 5 7/8 in W x 9 1/4 in H, stainless steel
Catalog No. 50-754-1195 Supplier USA Scientific Inc Supplier No. 26862415
24-box (3 long x 4 high) upright freezer rack for 50-place boxes, 19 in L x 5 7/8 in W x 9 1/4 in H, stainless steel