Learn More
TWD Scientific LLC 24 Position Microtitier SBS Format Rack. Pack of 10.
SDP
Catalog No. NC9786740 Shop All TWD Scientific LLC Products
Encompass_Preferred
24-position microtitier footprint rack for 1 dram vials (4ml, 15x45mm) & any vials with 15mm OD. Automation Friendly. SBS Format. 10/Pack
Catalog No. NC9786740 Supplier TWD Scientific LLC Supplier No. 5002002
24-position microtitier footprint rack for 1 dram vials (4ml, 15x45mm) & any vials with 15mm OD. Automation Friendly. SBS Format. 10/Pack