Learn More
Organomation Unheated 24 Position N-EVAP Nitrogen Evaporator
SDP
Catalog No. NC9184239
Encompass_Preferred
The unheated 24 position N-EVAP is our most adaptable benchtop evaporator. The 24 position N-EVAP offers the most customizable options for your sample preparation needs. The standard sample holder can accommodate samples with an outside diameter of 10-30 mm. However, with the optional 8 position large sample holder, vials with an outside diameter of 30-70 mm can be processed.These economical units provide controlled concentration of sample vials through the application of nitrogen gas and ambient temperatures for heat sensitive applications. The dual band spring hoist assembly provides a simple and effective means of removing samples from the heated dry bath.
Catalog No. NC9184239 Supplier Organomation Supplier No. 11250O
The unheated 24 position N-EVAP is our most adaptable benchtop evaporator. The 24 position N-EVAP offers the most customizable options for your sample preparation needs. The standard sample holder can accommodate samples with an outside diameter of 10-30 mm. However, with the optional 8 position large sample holder, vials with an outside diameter of 30-70 mm can be processed.These economical units provide controlled concentration of sample vials through the application of nitrogen gas and ambient temperatures for heat sensitive applications. The dual band spring hoist assembly provides a simple and effective means of removing samples from the heated dry bath.