Learn More
Enzo Life Sciences 24(S),25-Epoxycholesterol (5mg). CAS: 77058-74-3
SDP
Supplier:  Enzo Life Sciences BMLGR2310005
Encompass_Preferred
Endogenous agonist for the nuclear receptor LXR. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (10mg/ml) or DMSO (10mg/ml - requires warming to ~40°C which has been shown to lead to significant decomposition of the product). Long Term Storage: -20°C.
Catalog No. 50-201-0938