Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 24(S)-hydroxycholesterol | 474-73-7 | MFCD03695560 | 402.65 | C27H46O2 | 10 MG
SDP

Catalog No. 5000299244
Encompass_Preferred

24(S)-Hydroxycholesterol is an endogenous brain cholesterol metabolite supplied for research use. It functions as a potent liver X receptor (LXR) agonist and a selective positive allosteric modulator of N-methyl-D-aspartate receptors (NMDARs). The compound is provided as a powder and requires cold storage.

  • Endogenous brain cholesterol metabolite.
  • Potent liver X receptor (LXR) agonist.
  • Selective positive allosteric modulator of N-methyl-D-aspartate receptors (NMDARs).
  • Supplied as a powder for laboratory use.
  • Molecular formula C27H46O2; molecular weight 402.65.
  • CAS number 474-73-7.
  • Recommended storage: powder at -20°C; in solvent, -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-99244 Supplier Medchemexpress LLC Supplier No. HY1694010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

24(S)-Hydroxycholesterol is an endogenous brain cholesterol metabolite supplied for research use. It functions as a potent liver X receptor (LXR) agonist and a selective positive allosteric modulator of N-methyl-D-aspartate receptors (NMDARs). The compound is provided as a powder and requires cold storage.

  • Endogenous brain cholesterol metabolite.
  • Potent liver X receptor (LXR) agonist.
  • Selective positive allosteric modulator of N-methyl-D-aspartate receptors (NMDARs).
  • Supplied as a powder for laboratory use.
  • Molecular formula C27H46O2; molecular weight 402.65.
  • CAS number 474-73-7.
  • Recommended storage: powder at -20°C; in solvent, -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.