Learn More
Electron Microscopy Sciences 24 Place Scintillation Vial Rack with Handle
SDP
Catalog No. 50949346
Encompass_Preferred
Holds 24 scintillation vials (30mm diameter), with a handle on both ends for carrying, autoclavable. Measures: 298x127x95mm high, rugged and stackable.
Catalog No. 50-949-346 Supplier Electron Microscopy Sciences Supplier No. 72637
Holds 24 scintillation vials (30mm diameter), with a handle on both ends for carrying, autoclavable. Measures: 298x127x95mm high, rugged and stackable.