Learn More
Macherey-Nagel MN Square-well Block, 24 blocks
Catalog No. NC1420336
Encompass_Preferred
MN Square-well Block (24) 96-well blocks with 2.1 mL square wells pack of 24
Catalog No. NC1420336 Supplier Macherey-Nagel Supplier No. 740476.24
MN Square-well Block (24) 96-well blocks with 2.1 mL square wells pack of 24