Learn More
Chemglass Life Sciences 24-Well Cell Culture Plate, Treated, Flat, Sterile
SDP
Catalog No. 501897620
Encompass
• Made from high clarity, 100% virgin polystyrene• Sterilized by E-beam• Vacuum Plasma tissue culture treatment• Clear lot number for batch traceability• Peel-to-open, individually wrapped• Non-pyrogenic, DNase- and RNase-free
Catalog No. 50-189-7620 Supplier Chemglass Life Sciences Supplier No. CGE3503024
• Made from high clarity, 100% virgin polystyrene• Sterilized by E-beam• Vacuum Plasma tissue culture treatment• Clear lot number for batch traceability• Peel-to-open, individually wrapped• Non-pyrogenic, DNase- and RNase-free