Learn More
Socal Biomedical 24 WELL CELL CULTURE PLATE
Catalog No. NC1990019
Encompass_Preferred
24 Well Cell Culture Plate, Flat Bottom, Non-Treated, sterile
Catalog No. NC1990019 Supplier Socal Biomedical Supplier No. 702011
Add to Cart
Add to Cart
24 Well Cell Culture Plate, Flat Bottom, Non-Treated, sterile