Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BIO-CERT™ CELL CULTURE QUALITY 2-in-1 Culture Inserts for 24-well Plates, 8 μL Pore Size, PET Membrane

Catalog No. 14381000 Shop All BrandTech Scientific Products
Change view
Click to view available options
Packaging:
Multi-Pack Blister, 48/Pk.
Single Blister, 48/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
14-381-000 Single Blister, 48/Pk.
14-381-001 Multi-Pack Blister, 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-381-000 Supplier BRANDTECH™ BRAND™ Supplier No. 782716
Add to Cart
Add to Cart

Sterile inserts for 24-well plates, featuring cellGrade™ plus surface treatment for uniform cell growth. Designed with feet for standing or a removable hook for hanging, offering versatile use in cell culture applications.

  • Suitable for 2D and 3D cell cultures
  • TC treated with cellGrade™ plus surface treatment for enhanced cell growth
  • Highly transparent PET membrane ideal for imaging
  • Sterile according to ISO 11137, SAL 10-6
  • Produced in a Class 8 cleanroom per ISO 14644-1
  • Non-cytotoxic, meeting ISO 10993-5 standards
  • BIO-CERT™ CELL CULTURE QUALITY means inserts are free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Diameter size of 8.8 mm for an ideal fit of 24-well plates
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Breakapart No
Certifications/Compliance Sterile according to ISO 11137, SAL 10-6, Non-cytotoxic according to ISO 10993-5, pyrogen and endotoxin free to 0.01EU/mL
Certified Yes
Lid Without Lid
Content And Storage Ambient
Culture Area 0.6 cm2
Membrane PET
Autoclavable Not Autoclavable
Packaging Single Blister, 48/Pk.
Pore Size 8 μm
Binding Type cellGrade plus
Color Clear
Disposable Single-use
Product Type Microplate Insert
For Use With (Equipment) 24-well plate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 24
Volume (Metric) Working 100 to 400 μL
Sterility Sterile
Surface Treatment cellGrade Plus
Well Shape Round, Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) 2D and 3D cell culture, co-culture, migration, chemotaxis, TEER assays, and air-lift culture
Quantity 48
RNase Free Yes
Well Color Clear
Diameter (Metric) Membrane 8.8 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.