Learn More
Bellco Glass, Inc. 24 Well Glass Bottom Cell Culture Plate,Φ 10mm,TC, sterile 1/pk, 10/cs
SDP
Supplier:  Bellco Glass, Inc. 451400264
Encompass_Preferred
24 Well Glass Bottom Cell Culture Plate,Φ 10mm,TC, sterile 1/pk, 10/cs. glass bottom series are applied in confocal microscope, high resolution microscope, differential interference contrast microscope, polarized light microscope and phase contrast microscope for cell observation. • Can be used for live cell observation.• Special bottom design for easy grip.• Round cover glass insert for good appearance.• Medical adhesive glue to guarantee non-cytotoxicity.• Sterilized by E-beam, SAL=10-6.• Made of high clarity, 100% virgin polystyrene and high clarity glass as bottoms.• High quality cover glass of standard thickness.
Catalog No. 50-233-0710