Learn More
Cellvis 24 Well glass bottom plate with high performance #1.5 cover glass, 20/cs
Catalog No. NC0397150
Encompass
24 well glass bottom plate with high performance #1.5 cover glass (0.170+/-0.005mm). Black frame, with lid. Individually packed
Catalog No. NC0397150 Supplier Cellvis Supplier No. P241.5HN
24 well glass bottom plate with high performance #1.5 cover glass (0.170+/-0.005mm). Black frame, with lid. Individually packed