Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 24 well insert 0.4um PET PK48 (cellQART)
Supplier:  Altenloh Brinck & Co US INC. 9321002
Encompass_Preferred
these cellQART Cell Culture Inserts come with PET membranes in translucent optics in 24-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for fast cell growth / barrier formation.
Catalog No. 50-267-3486