Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 24 well insert 5.0um PET PK48 (cellQART)
Supplier:  Altenloh Brinck & Co US INC. 9325002
Encompass_Preferred
These cellQART Cell Culture Inserts come with PET membranes in translucent optics in 24-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for cell invasion, cell motility and chemotaxis.
Catalog No. 50-267-3479