Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 24 well insert 5.0um PET PK48 (cellQART)
Catalog No. 502673478
Encompass_Preferred
These cellQART Cell Culture Inserts come with PET membranes in clear optics in 24-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates.
Catalog No. 50-267-3478 Supplier Altenloh Brinck & Co US INC. Supplier No. 9325012
These cellQART Cell Culture Inserts come with PET membranes in clear optics in 24-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates.