Learn More
MatTek Corporation (Sleeve of 1) 24-Well Plate, No. 0 Uncoated Coverslip, 13 mm Glass Diameter
Supplier:  MatTek Corporation P24G013F.S
Encompass
With a proven track record of quality and convenience, MatTek dishes provide the optics of glass in standard size microscopy dishes and plates. Cited in thousands of scientific publications, MatTek dishes are a long-standing favorite of lab teams worldwide for their availability, ease-of-use, and the brilliant images they produce.These Multiwell Plates from MatTek Life Sciences combine the convenience of traditional plastic Falcon well plates with the optical quality of glass, providing superior microscopic images. For most applications, No. 1.5 is the preferred coverslip thickness, especially for optimizing image quality with high numerical aperture objectives. All MatTek plates are sterilized with gamma-irradiation and suitable for long term tissue culture. Compare to Falcon , Corning , Cellvis productsRecommended For: - Live cell imaging - Fluorescence microscopy- Confocal microscopy- Phase contrast microscopy- Differential interference contrast microscopy
Catalog No. NC1747112