Learn More
Bellco Glass, Inc. 24 Well Non-treated Plate with
SDP
Catalog No. 502330610
Encompass_Preferred
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume0.5–1.5mLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes
Catalog No. 50-233-0610 Supplier Bellco Glass, Inc. Supplier No. 229524000
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume0.5–1.5mLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes