Learn More
Cellvis 24 WELL PLATE 1CS 20/CS
Catalog No. NC1713666
Encompass
24 Well Plate with #1.5 glass-like polymer coverslip bottom, tissue culture treated for better cell attachment than cover glass1 Case 20/Case
Catalog No. NC1713666 Supplier Cellvis Supplier No. P241.5P
24 Well Plate with #1.5 glass-like polymer coverslip bottom, tissue culture treated for better cell attachment than cover glass1 Case 20/Case