Learn More
Greiner Bio-One 24-well Polystyrene Microplate with Flat Bottom Wells
Shop All Greiner Bio-One Products
Description
- Low autofluorescence
- Condensation ring lids enable optimal gas exchange
- Single position lids prevent cross-contamination
Specifications
Specifications
| Bottom Shape | Flat |
| Color | Black |
| Material | Polystyrene |
| Shape | Chimney |
| Type | Cell Culture Microplate |
| Well Count | 24 |