Learn More
Ted Pella Inc PELCO Prep-Eze™ 24-wellplate Insert
SDP
Catalog No. NC9979302
Encompass
The rectangular polypropylene Wellplate Inserts have 6, 12, or 24 wells and will fit into the Corning Costar Tissue Culture Plates of 6/12/24 Wellplate design. A 420µm opening, polypropylene mesh is at the bottom of each of the insert wells. The Wellplate Inserts may only be chemically sterilized. The outside dimensions of the Wellplate Inserts are: 81.3mm W x 123.5mm L x 24mm H (3.2" W x 4.86" L x 0.88" H)
Catalog No. NC9979302 Supplier Ted Pella Inc Supplier No. 36172
The rectangular polypropylene Wellplate Inserts have 6, 12, or 24 wells and will fit into the Corning Costar Tissue Culture Plates of 6/12/24 Wellplate design. A 420µm opening, polypropylene mesh is at the bottom of each of the insert wells. The Wellplate Inserts may only be chemically sterilized. The outside dimensions of the Wellplate Inserts are: 81.3mm W x 123.5mm L x 24mm H (3.2" W x 4.86" L x 0.88" H)