Learn More
Bellco Glass, Inc. 24 Well Tissue Culture Plate
SDP
Catalog No. 502329302
Encompass_Preferred
CELLTREAT Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alphanumeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. 1 Well Plate has the same footprint as standard multiple well plates and is useful for high volume and robotic screening of DNA libraries and manual plating it is also suitable for dotblotting bacterial clones on 11.4 x 7.3cm 4.5 x 2.875in nylon membranes. 4 well plate is a smaller footprint option which offers well size similar to that of a 24 well plate but has expanded well spacing to reduce cross contamination. 384 Well Plates are ideal for automated applications colorimetric assays compound storage or small volume applications.Specifications Lids have a stacking feature for added stability when handling tall stacks of plates
Catalog No. 50-232-9302 Supplier Bellco Glass, Inc. Supplier No. 229123000
Add to Cart
Add to Cart
CELLTREAT Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alphanumeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. 1 Well Plate has the same footprint as standard multiple well plates and is useful for high volume and robotic screening of DNA libraries and manual plating it is also suitable for dotblotting bacterial clones on 11.4 x 7.3cm 4.5 x 2.875in nylon membranes. 4 well plate is a smaller footprint option which offers well size similar to that of a 24 well plate but has expanded well spacing to reduce cross contamination. 384 Well Plates are ideal for automated applications colorimetric assays compound storage or small volume applications.Specifications Lids have a stacking feature for added stability when handling tall stacks of plates