Learn More
Bellco Glass, Inc. 24 Well Tissue Culture Plate
SDP
Catalog No. 502329890
Encompass_Preferred
Glass bottom plates have the same great features as standard tissue culture plates with the added benefit of borosilicate glass bottom microwellsConvenient live cell viewing without needing to transferOptically clear borosilicate glass for exceptional viewingApplications include: cell culture and imaging, live cell observation and specialty microscopy such as confocal high resolution and contrastTraditional footprints cause minimal disruption in cell culture techniquesConstructed with noncytotoxic medical adhesiveRound cover glass also available for use with glass bottom platesGlass bottom is #1.5 thickness Pack DescriptionIndividualCase Quantity5SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPlate: PolystyreneSecondary MaterialBottom: GlassWorking Volume0.5-1.5mLTissue Culture TreatedYesBottom ShapeFlatCell Growth Area0.78cm2RNase/DNase FreeYes
Catalog No. 50-232-9890 Supplier Bellco Glass, Inc. Supplier No. 229125000
Glass bottom plates have the same great features as standard tissue culture plates with the added benefit of borosilicate glass bottom microwellsConvenient live cell viewing without needing to transferOptically clear borosilicate glass for exceptional viewingApplications include: cell culture and imaging, live cell observation and specialty microscopy such as confocal high resolution and contrastTraditional footprints cause minimal disruption in cell culture techniquesConstructed with noncytotoxic medical adhesiveRound cover glass also available for use with glass bottom platesGlass bottom is #1.5 thickness Pack DescriptionIndividualCase Quantity5SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPlate: PolystyreneSecondary MaterialBottom: GlassWorking Volume0.5-1.5mLTissue Culture TreatedYesBottom ShapeFlatCell Growth Area0.78cm2RNase/DNase FreeYes