Learn More
CELLTREAT Scientific Products 24 Well Non-treated Plate, Sterile
SDP
Catalog No. 50202146
Encompass_Preferred
CELLTREAT Scientific Products 24 Well Non-Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with working volume of 0.5-1.5mL. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.
Catalog No. 50-202-146 Supplier CELLTREAT Scientific Products Supplier No. 229524
CELLTREAT Scientific Products 24 Well Non-Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with working volume of 0.5-1.5mL. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.