Learn More
Z And Z Medical Inc 24" Wide Blanket Warmer Cabinet
SDP
Supplier:  Z And Z Medical Inc PTHC155G
Encompass
The PTHC155G is a single chamber blanket warmer ideal for hospital settings keeping blankets at the proper temperature. The double pane tempered self-closing glass door ensures your staff can quickly check inventory with a display that goes down to the floor. This unit has a right-hand door swing with a pro style handle for added convenience. No internal fan helps to reduce lint buildup for safer, more efficient operation. Additionally, without fans or moving parts, there is no noise produced for silent operation. The interior walls are constructed from stainless steel to ensure easy cleanup and long-lasting durability. Sized with a 24" footprint, this unit takes up little space but can accommodate up to 12.17 cu.ft of storage. Inside, the PTHC155G is designed to operate between 90º and 140ºF with a foamed-in-place cabinet to ensure efficient insulation. This unit is featured with a digital thermostat that includes a large LED temperature readout.
Catalog No. NC2919826
Add to Cart
Request a Quote