Learn More
Alkali Scientific 24 x 1.5/2.0ml Hermetically sealed rotor, each (for C2417 series)
SDP
Catalog No. 5000663328
Encompass_Preferred
The 24 x 1.5/2.0mL Hermetically Sealed Rotor is designed for use with C2417 Series centrifuges, providing secure and efficient centrifugation for laboratory applications requiring contamination control and high speed performance. This rotor holds up to 24 microtubes ranging from 1.5mL to 2.0mL, making it suitable for molecular biology, clinical testing, and routine laboratory workflows. The hermetically sealed design helps prevent sample leakage and minimizes aerosol contamination, ensuring safe operation during centrifugation. Manufactured from high strength, corrosion resistant materials, the rotor offers durability and consistent performance. It supports speeds up to 15,000 RPM and is autoclavable at 121°C for sterilization. Compact dimensions and balanced construction provide stable operation for reliable sample processing in demanding laboratory environments.
Catalog No. 50-006-63328 Supplier Alkali Scientific Supplier No. RC24172420H
The 24 x 1.5/2.0mL Hermetically Sealed Rotor is designed for use with C2417 Series centrifuges, providing secure and efficient centrifugation for laboratory applications requiring contamination control and high speed performance. This rotor holds up to 24 microtubes ranging from 1.5mL to 2.0mL, making it suitable for molecular biology, clinical testing, and routine laboratory workflows. The hermetically sealed design helps prevent sample leakage and minimizes aerosol contamination, ensuring safe operation during centrifugation. Manufactured from high strength, corrosion resistant materials, the rotor offers durability and consistent performance. It supports speeds up to 15,000 RPM and is autoclavable at 121°C for sterilization. Compact dimensions and balanced construction provide stable operation for reliable sample processing in demanding laboratory environments.