Learn More
Leica Biosystems 24 x 24 x 6 mm Metal Base Mold for tissue embedding
Catalog No. NC3865248
Encompass
This metal base mold is designed for histology tissue embedding to form paraffin blocks for sectioning. The 24 x 24 x 6 mm cavity accommodates small to medium specimens and works with standard embedding cassettes.
  • Dimensions: 24 x 24 x 6 mm.
  • Reusable stainless steel construction for reliable thermal conductivity.
  • Rounded internal corners allow easy removal of formed wax blocks.
  • Compatible with standard tissue embedding cassettes for specimen orientation.
  • Pack quantity: 10 units per pack.
Catalog No. NC3865248 Supplier Leica Biosystems Supplier No. 3803082
This metal base mold is designed for histology tissue embedding to form paraffin blocks for sectioning. The 24 x 24 x 6 mm cavity accommodates small to medium specimens and works with standard embedding cassettes.
  • Dimensions: 24 x 24 x 6 mm.
  • Reusable stainless steel construction for reliable thermal conductivity.
  • Rounded internal corners allow easy removal of formed wax blocks.
  • Compatible with standard tissue embedding cassettes for specimen orientation.
  • Pack quantity: 10 units per pack.