Learn More
LabScientific Rectangular Coverslips, 24 x 50 mm, #1.5, 60 pack
Supplier:  LabScientific 7816
Encompass
Rectangular cover glasses packed in hinged boxes. All outer boxes are vacuum tropical packed. Thicknesses #1 = 0.13-0.16mm. #1.5 = 0.16-0.19mm
Catalog No. NC1034527