Learn More
Savillex Corporation Savillex Corporation 240 mL Standard Jar, PFA Material, -200°C to 260°C Temp Range, 240 mL
SDP
Catalog No. NC1913222
Encompass
The Savillex Corporation 240 mL Standard Jar is a versatile laboratory container designed for secure storage and transfer of various scientific materials. Its seamless construction and wide-mouth opening facilitate easy handling and content removal. The jar is made from durable molded PFA, offering a temperature range of -200°C to 260°C, making it ideal for demanding applications. Its translucent finish and ribbed grip ensure convenient use, while compatibility with 70 mm closures adds to its functionality for laboratory needs.
Catalog No. NC1913222 Supplier Savillex Corporation Supplier No. 100024001
The Savillex Corporation 240 mL Standard Jar is a versatile laboratory container designed for secure storage and transfer of various scientific materials. Its seamless construction and wide-mouth opening facilitate easy handling and content removal. The jar is made from durable molded PFA, offering a temperature range of -200°C to 260°C, making it ideal for demanding applications. Its translucent finish and ribbed grip ensure convenient use, while compatibility with 70 mm closures adds to its functionality for laboratory needs.