Learn More
Bellco Glass, Inc. 240 ul 384 well Deep Well Plates V Bottom Square Well NonSterile 5/pk 50/cs
SDP
Catalog No. 502329223
Encompass_Preferred
240 ul 384 well Deep Well Plates V Bottom Square Well NonSterile 5/pk 50/cs
Catalog No. 50-232-9223 Supplier Bellco Glass, Inc. Supplier No. 451400647
240 ul 384 well Deep Well Plates V Bottom Square Well NonSterile 5/pk 50/cs