Learn More
Electron Microscopy Sciences Clarified Polypropylene Container with lid, 240 ml
SDP
Catalog No. NC1463363
Encompass_Preferred
You can have confidence that these containers are designed for the safe, leak-resistant transport and storage of fluids. Screw-top containers have knurled cap and base for easy handling. They are graduated and made of durable clarified polypropylene. Sizing include lids.
Diameter: 3½ in
Height: 2-1/2 in
Catalog No. NC1463363 Supplier Electron Microscopy Sciences Supplier No. 6423107
You can have confidence that these containers are designed for the safe, leak-resistant transport and storage of fluids. Screw-top containers have knurled cap and base for easy handling. They are graduated and made of durable clarified polypropylene. Sizing include lids.
Diameter: 3½ in
Height: 2-1/2 in