Learn More
MSC Loctite 135354 10 mL Bottle Blue Liquid Threadlocker Medium Strength, 300°F Max, Series 242
Catalog No. 501113986
Encompass
Loctite 135354 10 mL Bottle Blue Liquid Threadlocker Medium Strength, 300°F Max, Series 242
Catalog No. 50-111-3986 Supplier MSC Supplier No. 88545009
Loctite 135354 10 mL Bottle Blue Liquid Threadlocker Medium Strength, 300°F Max, Series 242