Learn More
Sigma Aldrich Fine Chemicals Biosciences 245-Trimethylthiazole 98 f100G
Catalog No. 502898432
Encompass
2 4 5-Trimethylthiazole is a sulfur-containing heterocyclic compound mainly found in meat products such as chicken broth cooked beef and roasted lamb fat.
Catalog No. 50-289-8432 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W332518100GK
Add to Cart
Add to Cart
2 4 5-Trimethylthiazole is a sulfur-containing heterocyclic compound mainly found in meat products such as chicken broth cooked beef and roasted lamb fat.