Learn More
Alkali Scientific 245mm Square BioAssay Petri Di
SDP
Catalog No. 503029017
Encompass_Preferred
The 245mm Square BioAssay Petri Dish with Gridlines is designed to support a variety of laboratory applications including microbial cultivation high-throughput screening and bioassays Its gridlined surface allows for easy colony counting precise sample organization and efficient documentation making it a valuable tool for microbiology pharmaceutical research and environmental testing Constructed from non-treated polystyrene this petri dish is ideal for applications where enhanced cell attachment is not required providing versatility across different experimental needs The large 245mm square format offers an expansive surface area making it particularly suitable for high-volume sample testing and parallel analysis Each dish is sterile and individually packed to ensure contamination-free results in laboratory settings The packaging is designed for convenience with five dishes per pack and 20 dishes per case making it easy to manage inventory and optimize laboratory workflows These
Catalog No. 50-302-9017 Supplier Alkali Scientific Supplier No. PDB0024
The 245mm Square BioAssay Petri Dish with Gridlines is designed to support a variety of laboratory applications including microbial cultivation high-throughput screening and bioassays Its gridlined surface allows for easy colony counting precise sample organization and efficient documentation making it a valuable tool for microbiology pharmaceutical research and environmental testing Constructed from non-treated polystyrene this petri dish is ideal for applications where enhanced cell attachment is not required providing versatility across different experimental needs The large 245mm square format offers an expansive surface area making it particularly suitable for high-volume sample testing and parallel analysis Each dish is sterile and individually packed to ensure contamination-free results in laboratory settings The packaging is designed for convenience with five dishes per pack and 20 dishes per case making it easy to manage inventory and optimize laboratory workflows These