Learn More
Fine Science Tools Reusable Feeding Needles, 24 Gauge, 25 mm
SDP
Catalog No. NC9247204
Encompass
Product: Reusable Feeding Needle - Straight/25mm/24G/1.25mm Tip Diameter/ Gauge: 24/ Tip Diameter: 1.25mm/ Shape: Straight/ Tip Shape: Round/ Alloy: Stainless Steel/ Length: 25mm/ Sterile: No/ Style: Reusable/ Unit
Catalog No. NC9247204 Supplier Fine Science Tools Supplier No. 1806124
Product: Reusable Feeding Needle - Straight/25mm/24G/1.25mm Tip Diameter/ Gauge: 24/ Tip Diameter: 1.25mm/ Shape: Straight/ Tip Shape: Round/ Alloy: Stainless Steel/ Length: 25mm/ Sterile: No/ Style: Reusable/ Unit