Learn More
Roboz Surgical 24G X 1IN FEEDIN G NEEDLE
SDP
Catalog No. NC0145682
Encompass
Reusable Feeding Needle; Stainless Steel; Straight; 24 Gauge; 1 in. Length; 1.25mm Ball Diameter
Catalog No. NC0145682 Supplier Roboz Surgical Supplier No. FN7900
Add to Cart
Add to Cart
Reusable Feeding Needle; Stainless Steel; Straight; 24 Gauge; 1 in. Length; 1.25mm Ball Diameter