Learn More
Felix Storch Inc ACCUCOLD 24IN ADA REFRIG UNDERCOUNTER
SDP
Catalog No. NC3737777
Encompass_Preferred
  • 24-inch wide, ADA-compliant design
  • 5.5 cu. ft. capacity
  • 32 in height, suitable for lower counter installations
  • Automatic defrost system for reduced maintenance
  • Hidden evaporator for seamless interior cleaning
  • Fully finished white cabinet
  • Stainless steel wrapped door
  • Factory-installed lock for security
  • One-piece seamless liner
  • Adjustable wire shelves
  • Interior light for visibility
  • Door shelves for storage convenience
  • Crisper drawer for fresh items
  • Adjustable dial thermostat
  • 100% CFC free refrigerant
  • Freestanding installation
  • 115 V AC/60 Hz power supply
: https://www.accucold.com/product/FF6LW7SSHVADA
Catalog No. NC3737777 Supplier Felix Storch Inc Supplier No. FF6LW7SSHVADA
  • 24-inch wide, ADA-compliant design
  • 5.5 cu. ft. capacity
  • 32 in height, suitable for lower counter installations
  • Automatic defrost system for reduced maintenance
  • Hidden evaporator for seamless interior cleaning
  • Fully finished white cabinet
  • Stainless steel wrapped door
  • Factory-installed lock for security
  • One-piece seamless liner
  • Adjustable wire shelves
  • Interior light for visibility
  • Door shelves for storage convenience
  • Crisper drawer for fresh items
  • Adjustable dial thermostat
  • 100% CFC free refrigerant
  • Freestanding installation
  • 115 V AC/60 Hz power supply
: https://www.accucold.com/product/FF6LW7SSHVADA