Learn More
Uline 24 inch poly bag sealer
Catalog No. NC0729135
Encompass
electric heat impulse bag sealer for sealing poly bags up to 24 inches wide using impulse wire element for quick sealing through poly film with adjustable temperature and seal timer controls for customizing seal settings to make bottom, top and side seals on poly bags for industrial packaging applications
Catalog No. NC0729135 Supplier Uline Supplier No. H1246
electric heat impulse bag sealer for sealing poly bags up to 24 inches wide using impulse wire element for quick sealing through poly film with adjustable temperature and seal timer controls for customizing seal settings to make bottom, top and side seals on poly bags for industrial packaging applications