Learn More
Felix Storch Inc ACCUCOLD 24inch Wide All Purpose Refrigerator
SDP
Catalog No. NC1857387
Encompass_Preferred
The FF7LW is a counter height all-refrigerator with a full 5.5 cu.ft. capacity inside a slim 24" footprint. Designed for freestanding use, this model has a white exterior and user-reversible door swing. A factory installed front lock offers convenient security: https://www.accucold.com/product/FF7LW
Catalog No. NC1857387 Supplier Felix Storch Inc Supplier No. FF7LW
The FF7LW is a counter height all-refrigerator with a full 5.5 cu.ft. capacity inside a slim 24" footprint. Designed for freestanding use, this model has a white exterior and user-reversible door swing. A factory installed front lock offers convenient security: https://www.accucold.com/product/FF7LW