Learn More
PATHOLOGY INNOVATIONS The Peters Precision Cryoembedding System Medium Well Bar Set comes with one 24 mm well bar, 4 small chucks, 2 over-chuck freezing blocks and 8 dispensing slides.
Catalog No. NC3370380
Encompass_Preferred
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The Medium Well Bar Set comes with one 24 mm well bar, 4 small chucks, 2 over-chuck freezing blocks and 8 dispensing slides. This system was designed by inventor, professor and author of frozen section literature Dr. Stephen Peters to give users more precision and control when doing a high volume of frozen section procedures. This set is made for outfitting labs that process a high volume of frozen sections with average sized specimen, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.
Catalog No. NC3370380 Supplier PATHOLOGY INNOVATIONS Supplier No. 112
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The Medium Well Bar Set comes with one 24 mm well bar, 4 small chucks, 2 over-chuck freezing blocks and 8 dispensing slides. This system was designed by inventor, professor and author of frozen section literature Dr. Stephen Peters to give users more precision and control when doing a high volume of frozen section procedures. This set is made for outfitting labs that process a high volume of frozen sections with average sized specimen, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.