Learn More
Uline 24 Ounce Cold Packs
Catalog No. NC9399736
Encompass
Reusable 24-ounce cold pack gel pouch, freeze before use, 4 mil thick, 8 x 6 x 1.25"""", gel is non-toxic
Catalog No. NC9399736 Supplier Uline Supplier No. S7890
Reusable 24-ounce cold pack gel pouch, freeze before use, 4 mil thick, 8 x 6 x 1.25"""", gel is non-toxic