Learn More
USA Scientific Inc 24-pl .5mL Thermal-Lok block
Catalog No. 502744883
Encompass
24-place aluminum block for 0.5 mL tubes, for Thermal-Lok dry bath. 3 W x 3.75 D x 2 H.
Catalog No. 50-274-4883 Supplier USA Scientific Inc Supplier No. 25240000
24-place aluminum block for 0.5 mL tubes, for Thermal-Lok dry bath. 3 W x 3.75 D x 2 H.