Learn More
Perkin Elmer US LLC 24-Position Rack for SPB 50-24, 50 mL
Catalog No. 502103856
Encompass_Preferred
Exclusive RackLock feature locks DigiTUBEs in position within the rack allowing for easy, one-handed closure of screw cap.
Catalog No. 50-210-3856 Supplier Perkin Elmer US LLC Supplier No. N9308070
Exclusive RackLock feature locks DigiTUBEs in position within the rack allowing for easy, one-handed closure of screw cap.