Learn More
Sigma Aldrich Fine Chemicals Biosciences 24R2425 Dihydroxyvitamin D
Catalog No. 502907557
Encompass
(24R)-24 25-Dihydroxyvitamin D3 is a metabolite of vitamin D3. It is known to increase bone volume and mechanical strength.(24R) 24 25-Dihydroxyvitamin D3-26 26 26 27 27 27-d6nbspis a deuterated metabolite of vitamin D3 wherein methyl protons of C-26 C-27 carbons are replaced by deuterium.
Catalog No. 50-290-7557 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8029131ML
(24R)-24 25-Dihydroxyvitamin D3 is a metabolite of vitamin D3. It is known to increase bone volume and mechanical strength.(24R) 24 25-Dihydroxyvitamin D3-26 26 26 27 27 27-d6nbspis a deuterated metabolite of vitamin D3 wherein methyl protons of C-26 C-27 carbons are replaced by deuterium.